Learn More
ABclonal Technology PE Rabbit anti-Human TLR3 mAb
SDP
Catalog No. 5000012369
Encompass_Preferred
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity TLRs are highly conserved from Drosophila to humans and share structural and functional similarities They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity The various TLRs exhibit different patterns of expression This receptor is most abundantly expressed in placenta and pancreas and is restricted to the dendritic subpopulation of the leukocytes It recognizes dsRNA associated with viral infection and induces the activation of NF-kappaB and the production of type I interferons It thus plays a role in host defense against multiple viruses
Catalog No. 50-000-12369 Supplier ABclonal Technology Supplier No. A26593500T
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity TLRs are highly conserved from Drosophila to humans and share structural and functional similarities They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity The various TLRs exhibit different patterns of expression This receptor is most abundantly expressed in placenta and pancreas and is restricted to the dendritic subpopulation of the leukocytes It recognizes dsRNA associated with viral infection and induces the activation of NF-kappaB and the production of type I interferons It thus plays a role in host defense against multiple viruses