Learn More
ABclonal Technology PE Rabbit anti-Human TMPRSS2
SDP
Catalog No. 5000009484
Encompass_Preferred
This gene encodes a protein that belongs to the serine protease family The encoded protein contains a type II transmembrane domain a receptor class A domain a scavenger receptor cysteine-rich domain and a protease domain Serine proteases are known to be involved in many physiological and pathological processes This gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage This protein also facilitates entry of viruses into host cells by proteolytically cleaving and activating viral envelope glycoproteins Viruses found to use this protein for cell entry include Influenza virus and the human coronaviruses HCoV-229E MERS-CoV SARS-CoV and SARS-CoV-2 (COVID-19 virus) Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-09484 Supplier ABclonal Technology Supplier No. A27410200T
This gene encodes a protein that belongs to the serine protease family The encoded protein contains a type II transmembrane domain a receptor class A domain a scavenger receptor cysteine-rich domain and a protease domain Serine proteases are known to be involved in many physiological and pathological processes This gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage This protein also facilitates entry of viruses into host cells by proteolytically cleaving and activating viral envelope glycoproteins Viruses found to use this protein for cell entry include Influenza virus and the human coronaviruses HCoV-229E MERS-CoV SARS-CoV and SARS-CoV-2 (COVID-19 virus) Alternatively spliced transcript variants encoding different isoforms have been found for this gene