Learn More
ABclonal Technology PE Rabbit anti-Human TNF-a mAb
SDP
Catalog No. 5000010591
Encompass_Preferred
This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily This cytokine is mainly secreted by macrophages It can bind to and thus functions through its receptors TNFRSF1A TNFR1 and TNFRSF1B TNFBR This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation differentiation apoptosis lipid metabolism and coagulation This cytokine has been implicated in a variety of diseases including autoimmune diseases insulin resistance psoriasis rheumatoid arthritis ankylosing spondylitis tuberculosis autosomal dominant polycystic kidney disease and cancer Mutations in this gene affect susceptibility to cerebral malaria septic shock and Alzheimer disease Knockout studies in mice also suggested the neuroprotective function of this cytokine
Catalog No. 50-000-10591 Supplier ABclonal Technology Supplier No. A27433100T
This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily This cytokine is mainly secreted by macrophages It can bind to and thus functions through its receptors TNFRSF1A TNFR1 and TNFRSF1B TNFBR This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation differentiation apoptosis lipid metabolism and coagulation This cytokine has been implicated in a variety of diseases including autoimmune diseases insulin resistance psoriasis rheumatoid arthritis ankylosing spondylitis tuberculosis autosomal dominant polycystic kidney disease and cancer Mutations in this gene affect susceptibility to cerebral malaria septic shock and Alzheimer disease Knockout studies in mice also suggested the neuroprotective function of this cytokine