Learn More
ABclonal Technology PE Rabbit anti-Human TROP-2
Supplier: ABclonal Technology A26262500T
This intronless gene encodes a carcinoma-associated antigen This antigen is a cell surface receptor that transduces calcium signals Mutations of this gene have been associated with gelatinous drop-like corneal dystrophy