Learn More
ABclonal Technology PE Rabbit anti-Mouse CD14 mAb
SDP
Catalog No. 5000010209
Encompass_Preferred
This gene encodes a protein that plays an important role in the innate immune response and is expressed in monocyte macrophage cells This gene product acts as a co-receptor that binds several microbial and fungal molecules including lipopolysaccharides (LPS) This proteins LPS-binding activity is enhanced by the LPS binding protein (LBP) to allow binding to the TLR4-MD-2 co-receptor complex The product of this gene is found in two forms either as a soluble protein or attached to the cell surface by a glycosylphosphatidylinositol anchor
Catalog No. 50-000-10209 Supplier ABclonal Technology Supplier No. A26053100T
This gene encodes a protein that plays an important role in the innate immune response and is expressed in monocyte macrophage cells This gene product acts as a co-receptor that binds several microbial and fungal molecules including lipopolysaccharides (LPS) This proteins LPS-binding activity is enhanced by the LPS binding protein (LBP) to allow binding to the TLR4-MD-2 co-receptor complex The product of this gene is found in two forms either as a soluble protein or attached to the cell surface by a glycosylphosphatidylinositol anchor