Learn More
ABclonal Technology PE Rabbit anti-Mouse CD19 mAb
SDP
Catalog No. 5000013023
Encompass_Preferred
Involved in several processes including B-1 B cell differentiation positive regulation of phosphatidylinositol 3-kinase activity and positive regulation of release of sequestered calcium ion into cytosol Acts upstream of or within B cell receptor signaling pathway Located in external side of plasma membrane Is integral component of plasma membrane Is expressed in liver and spleen Human ortholog(s) of this gene implicated in common variable immunodeficiency Orthologous to human CD19 (CD19 molecule)
Catalog No. 50-000-13023 Supplier ABclonal Technology Supplier No. A25265500T
Involved in several processes including B-1 B cell differentiation positive regulation of phosphatidylinositol 3-kinase activity and positive regulation of release of sequestered calcium ion into cytosol Acts upstream of or within B cell receptor signaling pathway Located in external side of plasma membrane Is integral component of plasma membrane Is expressed in liver and spleen Human ortholog(s) of this gene implicated in common variable immunodeficiency Orthologous to human CD19 (CD19 molecule)