Learn More
ABclonal Technology PE Rabbit anti-Mouse CD204 mAb
SDP
Catalog No. 5000010847
Encompass_Preferred
Enables amyloid-beta binding activity cargo receptor activity and low-density lipoprotein particle binding activity Involved in several processes including endocytosis positive regulation of cholesterol storage and positive regulation of macrophage derived foam cell differentiation Acts upstream of or within establishment of localization in cell and lipoprotein transport Located in plasma membrane Is expressed in several structures including alimentary system early conceptus limb neural tube and reproductive system Human ortholog(s) of this gene implicated in Barrett s esophagus arteriosclerosis and prostate cancer Orthologous to human MSR1 (macrophage scavenger receptor 1)
Catalog No. 50-000-10847 Supplier ABclonal Technology Supplier No. A27278200T
Enables amyloid-beta binding activity cargo receptor activity and low-density lipoprotein particle binding activity Involved in several processes including endocytosis positive regulation of cholesterol storage and positive regulation of macrophage derived foam cell differentiation Acts upstream of or within establishment of localization in cell and lipoprotein transport Located in plasma membrane Is expressed in several structures including alimentary system early conceptus limb neural tube and reproductive system Human ortholog(s) of this gene implicated in Barrett s esophagus arteriosclerosis and prostate cancer Orthologous to human MSR1 (macrophage scavenger receptor 1)