Learn More
ABclonal Technology PE Rabbit anti-Mouse CD40 mAb
SDP
Catalog No. 5000011362
Encompass_Preferred
Predicted to enable antigen binding activity protein domain specific binding activity and ubiquitin protein ligase binding activity Involved in B cell mediated immunity CD40 signaling pathway and cellular calcium ion homeostasis Acts upstream of or within several processes including defense response to other organism positive regulation of B cell activation and positive regulation of interleukin-12 production Located in external side of plasma membrane and intracellular membrane-bounded organelle Part of CD40 receptor complex Is expressed in several structures including alimentary system brain hemolymphoid system gland liver and biliary system and reproductive system Human ortholog(s) of this gene implicated in several diseases including Kawasaki disease autoimmune disease (multiple) end stage renal disease hyperimmunoglobulin syndrome (multiple) and non-Hodgkin lymphoma (multiple) Orthologous to human CD40 (CD40 molecule)
Catalog No. 50-000-11362 Supplier ABclonal Technology Supplier No. A27174100T
Predicted to enable antigen binding activity protein domain specific binding activity and ubiquitin protein ligase binding activity Involved in B cell mediated immunity CD40 signaling pathway and cellular calcium ion homeostasis Acts upstream of or within several processes including defense response to other organism positive regulation of B cell activation and positive regulation of interleukin-12 production Located in external side of plasma membrane and intracellular membrane-bounded organelle Part of CD40 receptor complex Is expressed in several structures including alimentary system brain hemolymphoid system gland liver and biliary system and reproductive system Human ortholog(s) of this gene implicated in several diseases including Kawasaki disease autoimmune disease (multiple) end stage renal disease hyperimmunoglobulin syndrome (multiple) and non-Hodgkin lymphoma (multiple) Orthologous to human CD40 (CD40 molecule)