Learn More
ABclonal Technology PE Rabbit anti-Mouse CD44 mAb
SDP
Supplier:  ABclonal Technology A26063100T
Encompass_Preferred
This gene enables hyaluronic acid and type II TGF-beta receptor binding contributing to cytokine binding and receptor activity It regulates T cell activation protein phosphorylation and intracellular signaling Involved in Wnt signaling epithelium morphogenesis and wound healing it plays a role in inflammatory response Located in the basolateral plasma membrane external plasma membrane and microvilli it is part of the macrophage migration inhibitory factor receptor complex Expressed in various systems including alimentary branchial arch CNS genitourinary and limb Human orthologs are linked to breast carcinoma multiple carcinomas and prostate cancer It is orthologous to human CD44 a molecule in the Indian blood group
Catalog No. 50-000-12644