Learn More
ABclonal Technology PE Rabbit anti-Mouse CD6 mAb
SDP
Catalog No. 5000009938
Encompass_Preferred
Predicted to enable several functions including identical protein binding activity lipopolysaccharide binding activity and lipoteichoic acid binding activity Predicted to be involved in several processes including heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules lipopolysaccharide-mediated signaling pathway and positive regulation of macromolecule metabolic process Predicted to act upstream of or within cell adhesion Predicted to be located in immunological synapse Predicted to be spanning component of plasma membrane Predicted to be part of T cell receptor complex Predicted to be active in external side of plasma membrane Orthologous to human CD6 (CD6 molecule)
Catalog No. 50-000-09938 Supplier ABclonal Technology Supplier No. A27330100UG
Predicted to enable several functions including identical protein binding activity lipopolysaccharide binding activity and lipoteichoic acid binding activity Predicted to be involved in several processes including heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules lipopolysaccharide-mediated signaling pathway and positive regulation of macromolecule metabolic process Predicted to act upstream of or within cell adhesion Predicted to be located in immunological synapse Predicted to be spanning component of plasma membrane Predicted to be part of T cell receptor complex Predicted to be active in external side of plasma membrane Orthologous to human CD6 (CD6 molecule)