Learn More
ABclonal Technology PE Rabbit anti-Mouse CD62L mAb
SDP
Catalog No. 5000011194
Encompass_Preferred
Enables cell adhesion molecule binding activity Acts upstream of or within response to ATP Located in external side of plasma membrane Is expressed in brain brainstem submandibular gland epithelium and testis Used to study type 1 diabetes mellitus Human ortholog(s) of this gene implicated in Crohn s disease IgA glomerulonephritis and ulcerative colitis Orthologous to human SELL (selectin L)
Catalog No. 50-000-11194 Supplier ABclonal Technology Supplier No. A26715200T
Enables cell adhesion molecule binding activity Acts upstream of or within response to ATP Located in external side of plasma membrane Is expressed in brain brainstem submandibular gland epithelium and testis Used to study type 1 diabetes mellitus Human ortholog(s) of this gene implicated in Crohn s disease IgA glomerulonephritis and ulcerative colitis Orthologous to human SELL (selectin L)