Learn More
ABclonal Technology PE Rabbit anti-Mouse CD68 mAb
Supplier: ABclonal Technology A26390500T
Involved in several processes including cellular response to lipopolysaccharide cellular response to oxidised low-density lipoprotein particle stimulus and negative regulation of dendritic cell antigen processing and presentation Acts upstream of or within aging and cellular response to organic substance Located in lysosome and plasma membrane Is expressed in several structures including central nervous system embryo mesenchyme heart blood vessel liver and spleen Orthologous to human CD68 (CD68 molecule)