Learn More
ABclonal Technology PE Rabbit anti-Mouse CD87 mAb
SDP
Supplier:  ABclonal Technology A26599200T
Encompass_Preferred
Predicted to enable enzyme binding activity protein domain specific binding activity and signaling receptor binding activity Predicted to be involved in several processes including negative regulation of apoptotic signaling pathway positive regulation of epidermal growth factor receptor signaling pathway and positive regulation of release of cytochrome c from mitochondria Is integral component of plasma membrane Is expressed in several structures including decidua early conceptus and spleen Human ortholog(s) of this gene implicated in rheumatoid arthritis Orthologous to human PLAUR (plasminogen activator urokinase receptor)
Catalog No. 50-000-11919