Learn More
ABclonal Technology PE Rabbit anti-Mouse FOXP3 mAb
SDP
Catalog No. 5000008767
Encompass_Preferred
The protein encoded by this gene is a member of the forkhead winged-helix family of transcriptional regulators Defects in this gene result in the scurfy phenotype (sf) Alternative splicing results in multiple transcript variants
Catalog No. 50-000-08767 Supplier ABclonal Technology Supplier No. A26227200T
The protein encoded by this gene is a member of the forkhead winged-helix family of transcriptional regulators Defects in this gene result in the scurfy phenotype (sf) Alternative splicing results in multiple transcript variants