Learn More
ABclonal Technology PE Rabbit anti-Mouse IgM mAb
SDP
Catalog No. 5000009319
Encompass_Preferred
Enables several functions including antigen binding activity identical protein binding activity and immunoglobulin receptor binding activity Acts upstream of or within several processes including early endosome to late endosome transport immunoglobulin mediated immune response and positive regulation of B cell proliferation Located in external side of plasma membrane extracellular space and perinuclear region of cytoplasm Part of B cell receptor complex and immunoglobulin complex circulating Is expressed in several structures including brain hemolymphoid system and liver Used to study non-Hodgkin lymphoma and type 1 diabetes mellitus Human ortholog(s) of this gene implicated in agammaglobulinemia Orthologous to human IGHM (immunoglobulin heavy constant mu)
Catalog No. 50-000-09319 Supplier ABclonal Technology Supplier No. A26392200T
Enables several functions including antigen binding activity identical protein binding activity and immunoglobulin receptor binding activity Acts upstream of or within several processes including early endosome to late endosome transport immunoglobulin mediated immune response and positive regulation of B cell proliferation Located in external side of plasma membrane extracellular space and perinuclear region of cytoplasm Part of B cell receptor complex and immunoglobulin complex circulating Is expressed in several structures including brain hemolymphoid system and liver Used to study non-Hodgkin lymphoma and type 1 diabetes mellitus Human ortholog(s) of this gene implicated in agammaglobulinemia Orthologous to human IGHM (immunoglobulin heavy constant mu)