Learn More
ABclonal Technology PE Rabbit anti-Mouse IL-9 mAb
SDP
Catalog No. 5000010049
Encompass_Preferred
Enables cytokine activity Involved in positive regulation of interleukin-5 production Acts upstream of or within several processes including B cell activation regulation of cellular protein metabolic process and regulation of receptor signaling pathway via JAK-STAT Located in extracellular space Is expressed in gut liver thymus and thymus primordium Human ortholog(s) of this gene implicated in asthma and respiratory syncytial virus infectious disease Orthologous to human IL9 (interleukin 9)
Catalog No. 50-000-10049 Supplier ABclonal Technology Supplier No. A26265200T
Enables cytokine activity Involved in positive regulation of interleukin-5 production Acts upstream of or within several processes including B cell activation regulation of cellular protein metabolic process and regulation of receptor signaling pathway via JAK-STAT Located in extracellular space Is expressed in gut liver thymus and thymus primordium Human ortholog(s) of this gene implicated in asthma and respiratory syncytial virus infectious disease Orthologous to human IL9 (interleukin 9)