Learn More
Biolegend PE Rat IgG1, λ Isotype Ctrl
Supplier:  Biolegend 401905
Encompass_Preferred
PE Rat IgG1, λ Isotype Ctrl [G0114F7]; Isotype: Rat IgG1, λ; Reactivity: Rat; Apps: FC; Size: 25 ug
Catalog No. 50-605-421