Learn More
Biolegend PE Rat IgM, κ Isotype Ctrl
Supplier:  Biolegend 400808
Encompass_Preferred
PE Rat IgM, κ Isotype Ctrl [RTK2118]; Isotype: Rat IgM, κ; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-168-982