Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc PE Rt IgG2b Iso Con Anti Isoty

Catalog No. 5000086898
Encompass_Preferred

Product Name: PE Rat IgG2b Isotype Control (LTF-2)
Product Class: Antibody
Type: Isotype Control
Product Name: PE Rat IgG2b Isotype Control (LTF-2)
Cytek Catalog Number: 50-4031-U100
Marker: Isotype Control
Clonality: Monoclonal
Clone: LTF-2
Host Species: Rat
Isotype: IgG2b
Conjugation: PE
Size: 100 μg

Catalog No. 50-000-86898 Supplier Cytek Biosciences Inc Supplier No. 504031U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: PE Rat IgG2b Isotype Control (LTF-2)
Product Class: Antibody
Type: Isotype Control
Product Name: PE Rat IgG2b Isotype Control (LTF-2)
Cytek Catalog Number: 50-4031-U100
Marker: Isotype Control
Clonality: Monoclonal
Clone: LTF-2
Host Species: Rat
Isotype: IgG2b
Conjugation: PE
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.