Learn More
International Crystal PE SAMPLE CARD 25/PK
Catalog No. NC9147286
Encompass_Preferred
Polyethylene IR Sample Cards, PTFE and polyethylene IR sample cards provide inexpensive disposable sample substrates for qualitative analysis, The 2 x 4 in. cards mount vertically in the universal slide mount in any spectrophotometer, 25/
Catalog No. NC9147286 Supplier International Crystal Supplier No. 00007365
Polyethylene IR Sample Cards, PTFE and polyethylene IR sample cards provide inexpensive disposable sample substrates for qualitative analysis, The 2 x 4 in. cards mount vertically in the universal slide mount in any spectrophotometer, 25/