Learn More
International Enviroguard™ PE 3209 Shoe Covers
Catalog No. 19472495 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Closure Type:
Elastic Ankle
Catalog No. Closure Type
19-472-495 Elastic Ankle
1 options
Catalog No. 19-472-495 Supplier International Enviroguard™ Supplier No. 3209
Add to Cart
Add to Cart
PE disposable Shoe Covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids.
PE disposable shoe covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids. More durable than polypropylene and typically used in a variety of environments to help protect against chemicals, dirt, dust, liquid splash, non-hazardous particulates, and even shallow immersion.
  • Water resistant and come with a strong elastic closure
  • One size fits most; fits up to a men's size 12 1/2 shoe
  • Can be worn in indoor and outdoor environments
  • Possible applications include healthcare, construction, pharmaceuticals, labs, movers, realtors, food processing, beauty industry, delivery services, and more.
Specifications
Size X-Large
Material Polyethylene
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Resistance Water
Product Type Controlled Environments Shoe Covers