Learn More
MSC PIP CoreShield 3.0 Cut Resistant Gloves, Size 11, ANSI A5, Nitrile Foam Coated, 1/Pair
Catalog No. 5000509278
Encompass
These cut-resistant gloves provide advanced protection and dexterity for demanding industrial tasks, including automotive assembly. Designed with an 18-gauge seamless knit construction, they feature a specialized fiber blend for superior cut resistance while maintaining comfort and flexibility. A durable nitrile micro-foam palm coating enhances grip in dry and slightly oily conditions.
  • Cut resistance rating of ANSI A5 and ISO E
  • Nitrile foam palm coating for improved tactile sensitivity and grip
  • Seamlessly knitted construction with reinforced thumb
  • Certified safe for food contact
  • Antistatic properties per EN 16350
  • Touchscreen compatible and machine washable
  • Fiberglass-free and OEKO-TEX 100 certified
  • Knit wrist design for a secure, comfortable fit
Catalog No. 50-005-09278 Supplier MSC Supplier No. 32786931
Add to Cart
Add to Cart
These cut-resistant gloves provide advanced protection and dexterity for demanding industrial tasks, including automotive assembly. Designed with an 18-gauge seamless knit construction, they feature a specialized fiber blend for superior cut resistance while maintaining comfort and flexibility. A durable nitrile micro-foam palm coating enhances grip in dry and slightly oily conditions.
  • Cut resistance rating of ANSI A5 and ISO E
  • Nitrile foam palm coating for improved tactile sensitivity and grip
  • Seamlessly knitted construction with reinforced thumb
  • Certified safe for food contact
  • Antistatic properties per EN 16350
  • Touchscreen compatible and machine washable
  • Fiberglass-free and OEKO-TEX 100 certified
  • Knit wrist design for a secure, comfortable fit