Learn More
MSC PIP CoreShield™ Cut-Resistant Gloves, 18-Gauge, Size S, White, 100/Case
Catalog No. 5000508887
Encompass
These CoreShield™ gloves are designed for high-performance hand protection in demanding industrial environments, featuring an 18-gauge, fiberglass-free liner for superior dexterity and tactile sensitivity. Engineered for comfort and durability, these gloves provide reliable cut, abrasion, and puncture resistance while ensuring a secure grip in dry and slightly oily conditions.
  • Provides A4 cut resistance for enhanced safety
  • Seamless construction improves comfort and breathability
  • Breathable foam nitrile coating disperses fluids on contact to improve grip
  • Reinforced area between thumb and forefinger extends service life
  • Touchscreen compatible and silicone free for versatile handling
  • Compliant with FDA 21 CFR 177.2600 for food contact
  • ESD safe and Oeko-Tex® standard 100 certified
Catalog No. 50-005-08887 Supplier MSC Supplier No. 37108404
These CoreShield™ gloves are designed for high-performance hand protection in demanding industrial environments, featuring an 18-gauge, fiberglass-free liner for superior dexterity and tactile sensitivity. Engineered for comfort and durability, these gloves provide reliable cut, abrasion, and puncture resistance while ensuring a secure grip in dry and slightly oily conditions.
  • Provides A4 cut resistance for enhanced safety
  • Seamless construction improves comfort and breathability
  • Breathable foam nitrile coating disperses fluids on contact to improve grip
  • Reinforced area between thumb and forefinger extends service life
  • Touchscreen compatible and silicone free for versatile handling
  • Compliant with FDA 21 CFR 177.2600 for food contact
  • ESD safe and Oeko-Tex® standard 100 certified