Learn More
MSC PIP Cut Resistant Gloves, HPPE, 18 Gauge, Foam Nitrile, 100/Pk.
Catalog No. 5000508315
Encompass
These cut-resistant gloves feature an 18-gauge, fiberglass-free HPPE liner designed for superior dexterity and tactile sensitivity. The seamless construction provides comfort and breathability, while the foam nitrile coating ensures excellent grip by dispersing fluids on contact. Engineered for safety and performance, these gloves include a reinforced thumb crotch to enhance durability in high-wear areas and are compatible with touchscreen devices.
  • Cut level A5 protection
  • ESD-safe construction suitable for static-sensitive environments
  • Compliant with FDA 21 CFR 177.2600 for food handling
  • Silicone-free for residue-free handling
  • Oeko-Tex® standard 100 certified to exclude harmful substances
  • Knit wrist prevents entry of debris
  • Reusable and launderable design
Catalog No. 50-005-08315 Supplier MSC Supplier No. 37110566
Add to Cart
Add to Cart
These cut-resistant gloves feature an 18-gauge, fiberglass-free HPPE liner designed for superior dexterity and tactile sensitivity. The seamless construction provides comfort and breathability, while the foam nitrile coating ensures excellent grip by dispersing fluids on contact. Engineered for safety and performance, these gloves include a reinforced thumb crotch to enhance durability in high-wear areas and are compatible with touchscreen devices.
  • Cut level A5 protection
  • ESD-safe construction suitable for static-sensitive environments
  • Compliant with FDA 21 CFR 177.2600 for food handling
  • Silicone-free for residue-free handling
  • Oeko-Tex® standard 100 certified to exclude harmful substances
  • Knit wrist prevents entry of debris
  • Reusable and launderable design