Learn More
MSC PIP CoreShield 3.0 Cut Resistant Gloves, 18 Gauge, Nitrile Foam, ANSI A5, Size 10, 1 Pr.
Catalog No. 5000509879
Encompass
These 18 gauge, cut-resistant gloves provide lightweight hand protection with an ergonomic design for precision tasks. Engineered with a seamlessly knitted liner composed of recycled polyester, PE, and steel fibers, they offer superior dexterity and comfort. The gloves feature a nitrile micro-foam palm coating for excellent grip in both dry and wet conditions, along with a reinforced thumb for added durability.
  • ANSI cut level A5 and ISO level E protection
  • Certified for food handling and compliant with OEKO-TEX 100
  • Fiberglass-free and silicone-free construction
  • Antistatic properties according to EN 16350
  • Touchscreen compatible for operational efficiency
  • Color-coded for easy size identification
  • Washable for extended use
  • Knit wrist cuff provides a secure fit
Catalog No. 50-005-09879 Supplier MSC Supplier No. 70463716
These 18 gauge, cut-resistant gloves provide lightweight hand protection with an ergonomic design for precision tasks. Engineered with a seamlessly knitted liner composed of recycled polyester, PE, and steel fibers, they offer superior dexterity and comfort. The gloves feature a nitrile micro-foam palm coating for excellent grip in both dry and wet conditions, along with a reinforced thumb for added durability.
  • ANSI cut level A5 and ISO level E protection
  • Certified for food handling and compliant with OEKO-TEX 100
  • Fiberglass-free and silicone-free construction
  • Antistatic properties according to EN 16350
  • Touchscreen compatible for operational efficiency
  • Color-coded for easy size identification
  • Washable for extended use
  • Knit wrist cuff provides a secure fit