Learn More
Chemglass Life Sciences Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-05
SDP
Catalog No. CG159805
Encompass_Preferred
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-05. Vented polyethylene stopper to minimize evaporation from Dewar flasks.
Catalog No. CG159805 Supplier Chemglass Life Sciences Supplier No. CG159805
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-05. Vented polyethylene stopper to minimize evaporation from Dewar flasks.