Learn More
MSC G-Tek CoreShield Seamless Knit Glove, HPPE, A6 Cut, 18 Gauge, Size 11, 100/Case
Catalog No. 5000508994
Encompass
These 18 gauge, A6 cut resistant gloves are engineered for high-level protection while maintaining tactile sensitivity and dexterity. Featuring a foam nitrile coating on the palm and fingers, they offer enhanced grip in dry and slightly oily environments. The fiberglass-free, seamless HPPE construction ensures comfort for extended wear, while specialized design elements support ESD-safe operations and touchscreen compatibility.
  • A6 cut protection level with ANSI puncture and abrasion level 3
  • Foam nitrile coating provides superior grip
  • Reinforced thumb crotch for increased durability
  • ESD safe construction with electrical properties below 108 Ω
  • Touchscreen compatible for modern device interaction
  • Seamless knit wrist prevents debris entry
  • Silicone-free and food certified under 21 CFR 177.2600
  • Oeko-Tex® standard 100 certified
Catalog No. 50-005-08994 Supplier MSC Supplier No. 37110780
These 18 gauge, A6 cut resistant gloves are engineered for high-level protection while maintaining tactile sensitivity and dexterity. Featuring a foam nitrile coating on the palm and fingers, they offer enhanced grip in dry and slightly oily environments. The fiberglass-free, seamless HPPE construction ensures comfort for extended wear, while specialized design elements support ESD-safe operations and touchscreen compatibility.
  • A6 cut protection level with ANSI puncture and abrasion level 3
  • Foam nitrile coating provides superior grip
  • Reinforced thumb crotch for increased durability
  • ESD safe construction with electrical properties below 108 Ω
  • Touchscreen compatible for modern device interaction
  • Seamless knit wrist prevents debris entry
  • Silicone-free and food certified under 21 CFR 177.2600
  • Oeko-Tex® standard 100 certified