Learn More
MSC PIP CoreShield 3.0 Cut-Resistant Gloves, Nitrile Palm, 18 Ga, Size Medium, 10/Pk.
Catalog No. 5000508622
Encompass
These CORESHIELD 3.0 cut-resistant gloves provide advanced hand protection for demanding work environments. Designed with an 18-gauge seamless knit, these gloves combine dexterity and durability with a high cut resistance rating. The palm is coated with black nitrile foam to enhance grip in dry, wet, and oily conditions, while the overall construction features anti-static and touchscreen-compatible properties.
  • ANSI/ISEA cut resistance level A8
  • ANSI abrasion level 3 and puncture level 3
  • Features foam nitrile palm coating for superior grip
  • Seamless liner made from high-performance polyethylene, recycled polyester, and spandex
  • Touchscreen compatible and anti-static construction
  • Reinforced thumb crotch for increased wear life
  • Reusable and launderable at 40°C
  • Length of 9.83 in. with 3 in. knit wrist cuff
Catalog No. 50-005-08622 Supplier MSC Supplier No. 37113602
Add to Cart
Add to Cart
These CORESHIELD 3.0 cut-resistant gloves provide advanced hand protection for demanding work environments. Designed with an 18-gauge seamless knit, these gloves combine dexterity and durability with a high cut resistance rating. The palm is coated with black nitrile foam to enhance grip in dry, wet, and oily conditions, while the overall construction features anti-static and touchscreen-compatible properties.
  • ANSI/ISEA cut resistance level A8
  • ANSI abrasion level 3 and puncture level 3
  • Features foam nitrile palm coating for superior grip
  • Seamless liner made from high-performance polyethylene, recycled polyester, and spandex
  • Touchscreen compatible and anti-static construction
  • Reinforced thumb crotch for increased wear life
  • Reusable and launderable at 40°C
  • Length of 9.83 in. with 3 in. knit wrist cuff