Learn More
MSC Ansell HyFlex® 11-757 Cut Resistant Gloves, Polyurethane, 18 Gauge, Size 6, 12 Pairs/Bag
Catalog No. 5000509315
Encompass
These 18-gauge cut-resistant gloves offer advanced protection and durability for demanding industrial environments. Featuring specialized fiber technology, they provide high-level defense against sharp objects while maintaining the dexterity and tactile sensitivity required for precision tasks.
  • Features INTERCEPT™ technology for high-performance cut resistance
  • Offers a 20% stronger barrier against cut hazards compared to similar gloves
  • Reinforced thumb crotch resists wear to extend service life
  • Touchscreen compatible design for operational convenience
  • Breathable materials and ergonomic contouring ensure comfort during extended use
  • Dermatologically tested and approved by Dermatest®
  • Suitable for dry and slightly oily applications
Catalog No. 50-005-09315 Supplier MSC Supplier No. 37112745
These 18-gauge cut-resistant gloves offer advanced protection and durability for demanding industrial environments. Featuring specialized fiber technology, they provide high-level defense against sharp objects while maintaining the dexterity and tactile sensitivity required for precision tasks.
  • Features INTERCEPT™ technology for high-performance cut resistance
  • Offers a 20% stronger barrier against cut hazards compared to similar gloves
  • Reinforced thumb crotch resists wear to extend service life
  • Touchscreen compatible design for operational convenience
  • Breathable materials and ergonomic contouring ensure comfort during extended use
  • Dermatologically tested and approved by Dermatest®
  • Suitable for dry and slightly oily applications