Learn More
MSC PIP G-Tek Cut Resistant Gloves, A9, 18-Gauge, 1 Pair
Catalog No. 5000508321
Encompass
These high-performance gloves provide A9 cut protection with a seamless knit construction designed for superior tactile sensitivity and dexterity. Featuring an 18-gauge HPPE liner and foam nitrile coating, they offer an ideal balance of durability, breathability, and high-level cut resistance, making them well-suited for demanding industrial environments.
  • A9 cut resistance per ASTM F2992-15
  • Foam nitrile coating provides enhanced grip, breathability, and durability
  • Reinforced thumb crotch for increased wear resistance
  • Touchscreen compatible and ESD safe construction
  • Complies with 21 CFR 177.2600 for food handling
  • Knit wrist design keeps out debris
  • Silicone-free and Oeko-Tex® standard 100 certified
  • Reusable and launderable at 40°C
Catalog No. 50-005-08321 Supplier MSC Supplier No. 37115128
These high-performance gloves provide A9 cut protection with a seamless knit construction designed for superior tactile sensitivity and dexterity. Featuring an 18-gauge HPPE liner and foam nitrile coating, they offer an ideal balance of durability, breathability, and high-level cut resistance, making them well-suited for demanding industrial environments.
  • A9 cut resistance per ASTM F2992-15
  • Foam nitrile coating provides enhanced grip, breathability, and durability
  • Reinforced thumb crotch for increased wear resistance
  • Touchscreen compatible and ESD safe construction
  • Complies with 21 CFR 177.2600 for food handling
  • Knit wrist design keeps out debris
  • Silicone-free and Oeko-Tex® standard 100 certified
  • Reusable and launderable at 40°C