Learn More
Scientific Commodities Inc MEDICAL GRADE PTFE MICRO TUBING (Cat.# BB311-22-100′ Roll)
Catalog No. NC1727278
Encompass_Preferred
MEDICAL GRADE PTFE MICRO TUBINGThis medical grade PTFE micro size tubing absorbs practically no common acids, bases or solvents. Do not confuse this grade of tubing with general purpose, commercial & industrial grade PTFE tubing; although it can be used in those applications. It is manufactured with a low level of extractables and is chemically inert. It can be safely used from -410°F (-277°C) TO 500°F (260°C). This translucent 58 shore D tubing is USP class VI certified. It also meets military spec #MIL-L-22129, UL-224. Do not use with molten alkali metals, fluorine or strong fluorinating agents. It can easily be autoclaved or eto sterilized.SCI Cat.# BB311-22Gauge Size: 22Dimensions: I.D. x O.D.: 0.028″ x 0.048″ / 0.711mm x 1.22mm
Catalog No. NC1727278 Supplier Scientific Commodities Inc Supplier No. BB31122100
MEDICAL GRADE PTFE MICRO TUBINGThis medical grade PTFE micro size tubing absorbs practically no common acids, bases or solvents. Do not confuse this grade of tubing with general purpose, commercial & industrial grade PTFE tubing; although it can be used in those applications. It is manufactured with a low level of extractables and is chemically inert. It can be safely used from -410°F (-277°C) TO 500°F (260°C). This translucent 58 shore D tubing is USP class VI certified. It also meets military spec #MIL-L-22129, UL-224. Do not use with molten alkali metals, fluorine or strong fluorinating agents. It can easily be autoclaved or eto sterilized.SCI Cat.# BB311-22Gauge Size: 22Dimensions: I.D. x O.D.: 0.028″ x 0.048″ / 0.711mm x 1.22mm