Learn More
Scientific Commodities Inc PE TUBING 0.38 X 1.09MM 100FT
Catalog No. nc0923369
Encompass_Preferred
MEDICAL GRADE POLYETHYLENE MICRO TUBING This non-sterile rigid but flexible micro medical grade polyethylene tubing has excellent chemical and gas resistance. It works extremely well in applications with vacuum or pressure. It can be sterilized with ethylene oxide or electron beam. This milky white translucent tubing is non-radiopaque and is non-reactive to tissue. Will not impart taste and odor. Meets FDA 21 cfr 177.1520 (c) and USP class VI requirements. The hardness is 95 SHORE A. This PE tubing does not contain plasticizers. It has a temperature range up to 215F Deg. (103 Deg.C). Tolerance is +/-.002 on PE/01 to PE/1. Tolerance is +/-.003 on PE/2 to PE/9. Tolerance is +/-.005 on PE/10 to PE/17. Manufactured in the United States.
Catalog No. NC0923369 Supplier Scientific Commodities Inc Supplier No. BB31695PE/2
MEDICAL GRADE POLYETHYLENE MICRO TUBING This non-sterile rigid but flexible micro medical grade polyethylene tubing has excellent chemical and gas resistance. It works extremely well in applications with vacuum or pressure. It can be sterilized with ethylene oxide or electron beam. This milky white translucent tubing is non-radiopaque and is non-reactive to tissue. Will not impart taste and odor. Meets FDA 21 cfr 177.1520 (c) and USP class VI requirements. The hardness is 95 SHORE A. This PE tubing does not contain plasticizers. It has a temperature range up to 215F Deg. (103 Deg.C). Tolerance is +/-.002 on PE/01 to PE/1. Tolerance is +/-.003 on PE/2 to PE/9. Tolerance is +/-.005 on PE/10 to PE/17. Manufactured in the United States.