Learn More
Scientific Commodities Inc PE Tubing | 1.57x2.08MM | 100ft
Catalog No. NC1586272
Encompass_Preferred
This non-sterile rigid but flexible micro medical grade polyethylene tubing has excellent chemical and gas resistance.  It works extremely well in applications with vacuum or pressure.  It can be sterilized with ethylene oxide or electron beam.  This milky white translucent tubing is non-radiopaque and is non-reactive to tissue.  Will not impart taste and odor.
Catalog No. NC1586272 Supplier Scientific Commodities Inc Supplier No. BB31695PE/10
This non-sterile rigid but flexible micro medical grade polyethylene tubing has excellent chemical and gas resistance.  It works extremely well in applications with vacuum or pressure.  It can be sterilized with ethylene oxide or electron beam.  This milky white translucent tubing is non-radiopaque and is non-reactive to tissue.  Will not impart taste and odor.