Learn More
Elemental Scientific Inc PE VIAL 50ML PRECLEAN 500/CS
SDP
Catalog No. NC0712110
Encompass
50mL Polypropylene with screw cap, conical (28mm OD) (500 each)
Catalog No. NC0712110 Supplier Elemental Scientific Inc Supplier No. V280260D
50mL Polypropylene with screw cap, conical (28mm OD) (500 each)