Learn More
Braintree Scientific PE10 - Polyethylene .011"""" x .024"""" per ft.,200 ft. Continuo
SDP
Catalog No. 501955495
Encompass
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**
Catalog No. 50-195-5495 Supplier Braintree Scientific Supplier No. PE10200FT
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**