Learn More
Biolegend PE/Dazzle™ 594 anti-human CD146
Supplier:  Biolegend 361026
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD146 [P1H12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1704190