Learn More
Teknova PEA 5%SHEEP BLOOD 20PK
SDP
Supplier:  Teknova P1063
Encompass
PEA AGAR PLATES WITH 5% SHEEP BLOOD. Ready to use PEA (phenylethyl alcohol) agar plates for the for the selective isolation of Gram positive bacteria including Listeria, Streptococcus, Staphylococcus spp. Must purchase a minimum quantity of 8.
Catalog No. NC1837138
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.