Learn More
Teknova PEA Agar Plates. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387644
Encompass
1.50% Tryptone, 0.50% Animal-Free Soytone,0.50% Sodium Chloride, 0.25% Phenylethyl Alcohol,1.50% Agar
Catalog No. 50-238-7644 Supplier Teknova Supplier No. P01604PK
1.50% Tryptone, 0.50% Animal-Free Soytone,0.50% Sodium Chloride, 0.25% Phenylethyl Alcohol,1.50% Agar