Learn More
Teknova PEA Agar Plates with 5% Sheeps Blood. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387739
Encompass
1.50% Tryptone, 0.50% Soy Peptone, 0.50% Sodium Chloride, 1.50% Agar, 0.25% Phenylethyl Alcohol, 5.00% Defibrinated Sheep's Blood
Catalog No. 50-238-7739 Supplier Teknova Supplier No. P10634PK
1.50% Tryptone, 0.50% Soy Peptone, 0.50% Sodium Chloride, 1.50% Agar, 0.25% Phenylethyl Alcohol, 5.00% Defibrinated Sheep's Blood