Learn More
Oxyrase Oxypras Pea Blood Agar 10/Pk
SDP
Supplier:  Oxyrase PPEABA
Encompass
Innovative and specifically made for anaerobes to improve recovery of injured cells. Arrive reduced and ready to use; stays reduced in air for hours. Saves tech time; helps eliminate errors. Store refrigerated for a extended shelf life. Meets Best Practice criteria for working with clinical anaerobes. This product consists of 10 individually wrapped plates
Catalog No. 50-200-5327