Learn More
Leica Biosystems PEACH CASSETTES 1000/CS
Catalog No. NC1721905
Encompass_Preferred
IP Biopsy II cassettes are one piece biopsy cassettes with 1mm pores and a 35 degree writing surface. The cassettes are offered taped for easy loading into a ink printer. Available in 7 colors in cases of 1000.
Catalog No. NC1721905 Supplier Leica Biosystems Supplier No. 38440505
IP Biopsy II cassettes are one piece biopsy cassettes with 1mm pores and a 35 degree writing surface. The cassettes are offered taped for easy loading into a ink printer. Available in 7 colors in cases of 1000.