Learn More
Biotium Peacock™ Prestained Protein Marker
Catalog No. 501964387
Encompass_Preferred
Peacock™ Prestained Protein Marker allows you to visually monitor protein separation during SDS-PAGE or protein transfer for western. It contains 8 blue bands from 10 kDa to 180 kDa, plus pink and green bands at 75 kDa and 25 kDa. Ready to load with no heating or other preparation needed.
Catalog No. 50-196-4387 Supplier Biotium Supplier No. 21530500UL
Peacock™ Prestained Protein Marker allows you to visually monitor protein separation during SDS-PAGE or protein transfer for western. It contains 8 blue bands from 10 kDa to 180 kDa, plus pink and green bands at 75 kDa and 25 kDa. Ready to load with no heating or other preparation needed.