Learn More
MSC Silver Needle Flash Fire Coverall, DuPont Nomex IIIA, 60 cal/cm², NFPA 2112, 1 ea
Catalog No. 5000516872
Encompass
The Silver Needle Flash Fire Coverall provides high-performance protection during confined-space, short-duration thermal events. Constructed with a DuPont Nomex IIIA outer shell and a Nomex/Lenzing quilted liner, this garment functions as a dual-barrier defense against extreme heat. The design incorporates a patented integrated harness system for a loose, insulating fit and fire-resistant closures to facilitate rapid removal in the event of thermal exposure.
  • ATPV of 60 cal/cm²
  • Exceeds NFPA 2112 standards and is UL certified
  • Integrated harness system sewn directly into the garment
  • Heavy-duty Nomex zippers with storm flaps
  • Nomex Velcro seals at the neck, wrists and ankles
  • Optional Kevlar lanyard storage in a built-in shoulder pouch
Catalog No. 50-005-16872 Supplier MSC Supplier No. 06547897
The Silver Needle Flash Fire Coverall provides high-performance protection during confined-space, short-duration thermal events. Constructed with a DuPont Nomex IIIA outer shell and a Nomex/Lenzing quilted liner, this garment functions as a dual-barrier defense against extreme heat. The design incorporates a patented integrated harness system for a loose, insulating fit and fire-resistant closures to facilitate rapid removal in the event of thermal exposure.
  • ATPV of 60 cal/cm²
  • Exceeds NFPA 2112 standards and is UL certified
  • Integrated harness system sewn directly into the garment
  • Heavy-duty Nomex zippers with storm flaps
  • Nomex Velcro seals at the neck, wrists and ankles
  • Optional Kevlar lanyard storage in a built-in shoulder pouch