Learn More
Labsource Inc Universal Peel Heat Seal, Heat Seal Technology, Resealable, Compatible with Multiple Plate Materials, 500 m x 115 mm
SDP
Catalog No. NC2061306
Encompass
Universal Peel Heat Seal is an innovative sealing solution for microplates, ensuring broad compatibility with various plate materials. This resealable, heat seal offers easy access through both peeling and needling methods, making it ideal for diverse scientific applications. With a roll size of 500 m x 115 mm, it provides efficient sealing for up to 6,250 seals, thereby enhancing laboratory workflows by offering a contaminant-free environment.
Catalog No. NC2061306 Supplier Labsource Inc Supplier No. P98570
Universal Peel Heat Seal is an innovative sealing solution for microplates, ensuring broad compatibility with various plate materials. This resealable, heat seal offers easy access through both peeling and needling methods, making it ideal for diverse scientific applications. With a roll size of 500 m x 115 mm, it provides efficient sealing for up to 6,250 seals, thereby enhancing laboratory workflows by offering a contaminant-free environment.