Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peanut Agglutinin (Biotinylated) | 1 MG
SDP

Catalog No. 5000426667
Encompass_Preferred

Peanut Agglutinin (Biotinylated) is a plant lectin used as a probe to specifically bind biomolecules like polysaccharides and peptides. This biological material is intended for life science research and is for research use only, not for sale to patients.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Intended for research use only
  • Activity: ≥85%
  • Biotinylation: ≥97%
  • Concentration: 5 mg/mL
  • Formulation: 10 mM HEPES, pH 7.5, 0.15 M NaCl, 0.08% sodium azide, 0.1 mM CaCl2, 0.01 mM MnCl2
  • Storage: Store at 4°C for 2 years, protect from light
  • Classified as a biochemical assay reagent
  • Classified as a native protein

Catalog No. 50-004-26667 Supplier Medchemexpress LLC Supplier No. HYNP01821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Peanut Agglutinin (Biotinylated) is a plant lectin used as a probe to specifically bind biomolecules like polysaccharides and peptides. This biological material is intended for life science research and is for research use only, not for sale to patients.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Intended for research use only
  • Activity: ≥85%
  • Biotinylation: ≥97%
  • Concentration: 5 mg/mL
  • Formulation: 10 mM HEPES, pH 7.5, 0.15 M NaCl, 0.08% sodium azide, 0.1 mM CaCl2, 0.01 mM MnCl2
  • Storage: Store at 4°C for 2 years, protect from light
  • Classified as a biochemical assay reagent
  • Classified as a native protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.