Learn More
MSC Proto Ratchet, 1/4 in Drive, Pear Head, 45-Tooth, 1 ea
Catalog No. 501920083
Encompass
This premium pear head ratchet features a 1/4 inch drive, designed for access in tight work areas. It incorporates a 45-tooth gear for balanced articulation and high torque, and an ergonomically designed handle for comfortable, slip-resistant operation.
  • Small pear head design for access in tight, hard-to-reach work areas
  • A 45-tooth gear provides balance between 8° articulation angle and high torque carrying capacity
  • Low profile reversing lever design for quick, one-hand operation
  • Contoured, ergonomically designed handle for comfort
  • Knurled bands allow for increased grip and slip resistance
  • Full polish finish offers double nickel chrome plating for enhanced corrosion protection
  • Field repairable
Catalog No. 50-192-0083 Supplier MSC Supplier No. 39974381
This premium pear head ratchet features a 1/4 inch drive, designed for access in tight work areas. It incorporates a 45-tooth gear for balanced articulation and high torque, and an ergonomically designed handle for comfortable, slip-resistant operation.
  • Small pear head design for access in tight, hard-to-reach work areas
  • A 45-tooth gear provides balance between 8° articulation angle and high torque carrying capacity
  • Low profile reversing lever design for quick, one-hand operation
  • Contoured, ergonomically designed handle for comfort
  • Knurled bands allow for increased grip and slip resistance
  • Full polish finish offers double nickel chrome plating for enhanced corrosion protection
  • Field repairable