Learn More
Biomatik Corporation Pear1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11279
Encompass_Preferred
Pear1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PEAR1. The Pear1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Platelet Endothelial Aggregation Receptor 1 Protein (932-1037Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PEAR1. Accession Number: Q5VY43. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9246