Learn More
Perkin Elmer US LLC Pearl Nebulizer Cleaner
Catalog No. 502104156
Encompass_Preferred
With the Pearl, a cleaning solution can be pumped manually in reverse, through both the sample and gas passages. When the wet-cleaning process is finished, rinse with DI water and flush dry with compressed air or inert gas.
Catalog No. 50-210-4156 Supplier Perkin Elmer US LLC Supplier No. N8145457
With the Pearl, a cleaning solution can be pumped manually in reverse, through both the sample and gas passages. When the wet-cleaning process is finished, rinse with DI water and flush dry with compressed air or inert gas.