Learn More
Meinhard Elemental Scientific Pearl Nebulizer Cleaner for all Quartz and Glass Concentric Nebulizers
Catalog No. NC1679066
Encompass
With this device a cleaning solution can be pumped manually in reverse, through both the sample and gas passages. Favorite cleaning solutions are - Sigma-Aldrich RBS-25 diluted about 20x, Deconex similarly diluted; - or - either of these, but with the addition of 10% methanol. When the wet-cleaning process is finished, rinse with DI water and flush dry with compressed air or inert gas.
Catalog No. NC1679066 Supplier Meinhard Elemental Scientific Supplier No. PRL03
With this device a cleaning solution can be pumped manually in reverse, through both the sample and gas passages. Favorite cleaning solutions are - Sigma-Aldrich RBS-25 diluted about 20x, Deconex similarly diluted; - or - either of these, but with the addition of 10% methanol. When the wet-cleaning process is finished, rinse with DI water and flush dry with compressed air or inert gas.