Learn More
Sigma Aldrich Fine Chemicals Biosciences PearlTM Pipette adjustable1EA
Catalog No. 502901502
Encompass
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI) as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Catalog No. 50-290-1502 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS1201041EA
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI) as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.